missing translation for 'onlineSavingsMsg'
Learn More

Docosane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014961
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014961 5 g
111014960 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014961 Supplier Merck Emd Millipore Supplier No. 439425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Docosane can find applications as a phase change material, known as latent heat storage media, which can help in storage and conversion of solar energy.

Petroleum

Specifications

CAS 629-97-0
Vapor Pressure <1 mmHg (21.1°C)
Vapor Density 10.8 (vs. air)
Density 0.778 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula CH3(CH2)20 CH3
MDL Number MFCD00009348
Melting Point 42°C to 45°C; 43°C to 46°C
Boiling Point 369°C
Type Analytical Standard
Quantity 5 g
Formula Weight 310.60
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less