missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docosane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Description
Petroleum
Specifications
Specifications
| CAS | 629-97-0 |
| Vapor Pressure | <1 mmHg (21.1°C) |
| Vapor Density | 10.8 (vs. air) |
| Density | 0.778 g/mL (at 25°C) |
| Flash Point | 113°C |
| Physical Form | Neat |
| Linear Formula | CH3(CH2)20 CH3 |
| MDL Number | MFCD00009348 |
| Melting Point | 42°C to 45°C; 43°C to 46°C |
| Boiling Point | 369°C |
| Show More |