missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docetaxel Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Docetaxel is a taxoid anticancer agent that binds to β tubulin and promotes tubulin assembly into microtubles inducing cell-cycle arrest and apoptosis.
Specifications
Specifications
| CAS | 148408-66-6 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C43H53 NO14 · 3H2 O |
| Synonym | Docetaxel trihydrate |
| Type | Certified Reference Material |
| Quantity | 200 mg |
| Formula Weight | 861.93 |
| Grade | Certified Reference Material |