missing translation for 'onlineSavingsMsg'
Learn More

Docetaxel, 99%

Docetaxel, CAS # 114977-28-5, is a cytotoxic, semisynthetic, second-generation taxane derived from the needles of the European yew tree and displays antineoplastic, antibiotic, and antimicrotubule activities. | CAS: 114977-28-5 | C43H53NO14 | 807.89 g/mol

Supplier:  Thermo Scientific Chemicals J60174MC

 View more versions of this product

Catalog No. AAJ60174MC

Only null left

Description

Description

Docetaxel is a second-generation cytotoxic antimicrotubule agent and is mostly used in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Docetaxel is an analog of taxol, and is considered to be a cytotoxic, semisynthetic, second-generation taxane that exhibits antineoplastic, antibiotic, and antimicrotubule activities
• Docetaxel is capable of inhibiting cellular mitosis. This compound promotes the assembly of microtubules from tubulin dimers and stabilizes microtubules by preventing depolymerization. This stability inhibits the normal dynamic reorganization of the microtubule network that is essential for vital interphase and mitotic cellular functions. This results in the persistence of abnormal microtubule structures leading to cell-cycle arrest and cell death

Application

• Docetaxel can inhibit angiogenesis by acting on vascular endothelial growth factor (VEGF) associated with cancer cells
• This compound has been used to investigate the effect of lysosomal autophagic flux in gastric cancer cells

Specifications

Specifications

Docetaxel
186°C to 192°C
C43H53NO14
100 mg
docetaxel, taxotere, docetaxel anhydrous, docetaxol, emdoc, docetaxel, trihydrate, docetaxel inn, taxotere tn, unii-699121phca, txl
ZDZOTLJHXYCWBA-VCVYQWHSSA-N
807.89
CHEBI:4672
99%
114977-28-5
White
MFCD00800737
14,3397
Soluble in dimethyl sulfoxide and ethanol; Insoluble in water.
CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1OC(=O)C(C(C5=CC=CC=C5)NC(=O)OC(C)(C)C)O)O)OC(=O)C6=CC=CC=C6)(CO4)OC(=O)C)O)C)O
148124
807.88
Powder

Chemical Identifiers

114977-28-5
807.89
ZDZOTLJHXYCWBA-VCVYQWHSSA-N
148124
CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1OC(=O)C(C(C5=CC=CC=C5)NC(=O)OC(C)(C)C)O)O)OC(=O)C6=CC=CC=C6)(CO4)OC(=O)C)O)C)O
C43H53NO14
MFCD00800737
docetaxel, taxotere, docetaxel anhydrous, docetaxol, emdoc, docetaxel, trihydrate, docetaxel inn, taxotere tn, unii-699121phca, txl
CHEBI:4672
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H361fd

RTECSNumber : DA4172750

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only