missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNP-X, SE, 6-(2,4-Dinitrophenyl)aminohexanoic Acid, Succinimidyl Ester

Catalog No. d2248
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2248 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2248 Supplier Invitrogen™ Supplier No. D2248

DNP-X, SE, 6-(2,4-Dinitrophenyl)aminohexanoic Acid, Succinimidyl Ester

The amine-reactive DNP-X succinimidyl ester can be used bioconjugates that can be detected with anti-dinitrophenyl (anti-DNP) antibodies. This reactive hapten contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate increase accessibility to anti-DNP antibodies.

Specifications

Quantity 25 mg
Product Type DNP-X Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye DNP
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.