missing translation for 'onlineSavingsMsg'
Learn More

5 Percent Ruthenium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical

Catalog No. 18607978
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18607978 100 g
18607976 5 g
18607977 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607978 Supplier Spectrum Chemical Mfg Cor Supplier No. SP401100G
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reduced 5% ruthenium catalyst supported on alumina powder for chemical synthesis applications

  • Free flowing powder with particle size of 27 microns
  • Purity characteristics and the strong metal support interaction which can be obtained with the metal

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Color Gray
Physical Form Powder or Liquid
Quantity 100 g
Solubility Information Insoluble in Water
Formula Weight 101.07 g/mol
Grade Not Available
Concentration or Composition (by Analyte or Components) 95% Aluminum Oxide, 5% Ruthenium
Packaging Amber Glass Bottle
Appearance Powder
Chemical Name or Material 5% Ruthenium on Alumina Powder, Reduced, Dry, 70 μ, BASF Catalyst

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.