missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJC12 Recombinant Rabbit Monoclonal Antibody (23GB1815)

Catalog No. MA553991
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553991 20 μL
MA553990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553991 Supplier Invitrogen™ Supplier No. MA553991
Only null left

Rabbit Recombinant Monoclonal Antibody

DNAJC12 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of a subclass of the HSP40/DnaJ protein family. Members of this family of proteins are associated with complex assembly, protein folding, and export. Two transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Antigen DNAJC12
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1815
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DNAJC12
Gene Accession No. Q9UKB3
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 12; DnaJ heat shock protein family (Hsp40) member C12; dnaJ homolog subfamily C member 12; DnaJ subfamily C member 12-like protein; DNAJC12; DnaJ-like protein subfamily C member 12-like protein; J domain containing protein 1; J domain containing protein 1 (JDP1); J domain protein 1; j domain-containing protein 1; JDP1; mJDP1
Gene Symbols DNAJC12
Host Species Rabbit
Immunogen A synthesized peptide derived from human DKC1 (140-198AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56521
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less