missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAJA4 Recombinant Rabbit Monoclonal Antibody (23GB5825)

Catalog No. MA552923
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552923 20 μL
MA552922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552923 Supplier Invitrogen™ Supplier No. MA552923
Only null left

Rabbit Recombinant Monoclonal Antibody

DNAJA4 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

DNAJA4 (DnaJ Heat Shock Protein Family (Hsp40) Member A4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include unfolded protein binding and heat shock protein binding.

Specifications

Antigen DNAJA4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5825
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DNAJA4
Gene Accession No. Q8WW22, Q9JMC3
Gene Alias 1110021L12Rik; AV358213; Dj4; DnaJ (Hsp40) homolog, subfamily A, member 4; DnaJ heat shock protein family (Hsp40) member A4; dnaJ homolog subfamily A member 4; DNAJA4; heat shock protein, DNAJ-like 4; Hsj4; mmDjA4; MST104; MSTP104; PRO1472
Gene Symbols DNAJA4
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJA4 (360-397AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300721, 55466, 58233
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less