missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAH14 Monoclonal Antibody (3C12)
GREENER_CHOICE

Catalog No. PIMA524515
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524515 Supplier Invitrogen™ Supplier No. MA524515
Only null left

Mouse Monoclonal Antibody

Peptide Sequence: MDKEETKTKP RLLRYEEKKY EDVKPLETQP AEIAEKETLE YKTVRTFSES LKSEKTEDYL RESIIQQHMV SPEPASLKEK GKSRRKKDQT HACPNVRKAR PVSYDRTEPK DDDVIRNIIR LREKLGWQTI LPQHSLKYGS SKIAIQKITL KKPLEDDGEF VYCLPRKSPK SLYNPYDLQV INIVGSVKE.

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP. Involved in sperm motility; implicated in sperm flagellar assembly.

Specifications

Antigen DNAH14
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene DNAH14
Gene Accession No. Q0VDD8
Gene Alias Axonemal beta dynein heavy chain 14; C1orf67; Ciliary dynein heavy chain 14; DNAH14; Dnahc14; dynein axonemal heavy chain 14; dynein heavy chain 14, axonemal; dynein, axonemal, heavy chain 14; dynein, axonemal, heavy polypeptide 14; HL18; HL-18
Gene Symbols DNAH14
Host Species Mouse
Immunogen MGC27277 (AAH42869, 1 a.a. ~ 189 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127602
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less