missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNAH11 Monoclonal Antibody (C2)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA566065

Catalog No. MA566065


Only null left
Explore available promotions

Description

Description

DNAH11 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a ciliary outer dynein arm protein and is a member of the dynein heavy chain family. It is a microtubule-dependent motor ATPase and has been reported to be involved in the movement of respiratory cilia. Mutations in this gene have been implicated in causing Kartagener Syndrome (a combination of situs inversus totalis and Primary Ciliary Dyskinesia (PCD), also called Immotile Cilia Syndrome 1 (ICS1)) and male sterility. [provided by RefSeq, Mar 2013].
Specifications

Specifications

DNAH11
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q96DT5
DNAH11
Recombinant Dynein, Axonemal, Heavy Chain 11 (DNAH11) (Leu3896-Asn4122).
200 μL
Primary
Human
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
C2
Unconjugated
DNAH11
axonemal beta dynein heavy chain 11; axonemal dynein heavy chain 11; CILD7; ciliary dynein heavy chain 11; DNAH11; DNAHBL; DNAHC11; DNHBL; DPL11; dynein axonemal heavy chain 11; dynein heavy chain 11, axonemal; dynein, axonemal, heavy chain 11; dynein, axonemal, heavy polypeptide 11; dynein, ciliary, heavy chain 11
Mouse
Protein A/G
RUO
8701
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions