Learn More
Applied Biosystems™ DNA Synthesis Column, Empty, 1.0 μmol crimp style
Catalog No. 400407
Encompass
Change view
Click to view available options
Quantity:
50 U
Catalog No. Quantity
400407 50 U
1 options
Catalog No. 400407 Supplier Applied Biosystems™ Supplier No. 400407
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Allows users to pack their own DNA systhesis columns.
  • Set of 50 empty columns
  • 1.0μmol crimp style
  • For use in nucleic acid labeling and oligo systhesis
Research Use Only. Not for use in diagnostics procedures.
Specifications
Packaging Type Shipped at Room Temperature
Quantity 50 U
For Research Use Only. Not for use in diagnostic procedures.