missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Retardation Gels (6%), 1.0 mm DFS Item

Novex™ DNA Retardation Gels are 6% polyacrylamide prepared with 0.5X TBE gel buffer, providing clear resolution of fragments used for DNA retardation assays from 60–2500 bp.

Supplier:  Invitrogen™ EC63652BOX

 View more versions of this product

Catalog No. EC63652BOX


Only null left
Explore available promotions

Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.

Description

Description

Novex™ DNA Retardation Gels are 6% polyacrylamide prepared with 0.5X TBE gel buffer, providing clear resolution of fragments used for DNA retardation assays from 60–2500 bp. This preparation is good for electrophoretic separation, yet low enough to promote DNA-protein interactions. Novex DNA Retardation Gels are designed to run on the XCell SureLock™ Mini-Cell and are available in 10-, 12-, or 15-well formats.

• DNA fragments ranging from 60-2,500 bp
• Prepared with 0.5X TBE as gel buffer
• Designed to run on the XCell SureLock Mini-Cell
• Available in 10, 12, and 15 well formats
• Manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS

To ensure best possible results, these gels are single use and individually wrapped. Detection is performed with ethidium bromide staining of DNA or, for greater sensitivity, with radiolabeling the DNA or protein.

Formulation
Novex DNA Retardation Gels are manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

• Catalog number refers to a single box of 10 gels

Store at 4°C.
6%
DNA Retardation
12-well
TBE Hi-Density Sample Buffer (5X)
Wet Ice
XCell SureLock Mini-Cell
DNA Retardation Gels, 12-well
1 mm
60 to 2500 bp
10 Gels/Box
Double-stranded DNA (dsDNA)
DNA Retardation Gel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions