missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNA Ligase IV Recombinant Rabbit Monoclonal Antibody (23GB2515)
Description
DNA Ligase IV Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.
Specifications
Specifications
| Antigen | DNA Ligase IV |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB2515 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | LIG4 |
| Gene Accession No. | P49917 |
| Gene Alias | 5830471N16Rik; DNA joinase; DNA ligase (ATP) 4; DNA ligase 4; DNA ligase IV; DNA repair enzyme; Lig4; LIG4S; ligase IV, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase; tiny |
| Gene Symbols | LIG4 |
| Show More |