missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Ligase IV Recombinant Rabbit Monoclonal Antibody (23GB2515)
GREENER_CHOICE

Catalog No. MA553494
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553494 100 μL
MA553495 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553494 Supplier Invitrogen™ Supplier No. MA553494
Only null left

Rabbit Recombinant Monoclonal Antibody

DNA Ligase IV Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.

Specifications

Antigen DNA Ligase IV
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2515
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LIG4
Gene Accession No. P49917
Gene Alias 5830471N16Rik; DNA joinase; DNA ligase (ATP) 4; DNA ligase 4; DNA ligase IV; DNA repair enzyme; Lig4; LIG4S; ligase IV, DNA, ATP-dependent; polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase; tiny
Gene Symbols LIG4
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNA Ligase IV (504-685AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3981
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less