missing translation for 'onlineSavingsMsg'
Learn More

DNA ligase I Mouse anti-Human, Mouse, Clone: 5H5, MilliporeSigma™

Catalog No. mabe190525
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE190525 25 μg
MABE1905100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE190525 Supplier MilliporeSigma Supplier No. MABE190525UG
Only null left

Mouse Monoclonal Antibody

DNA ligase I Monoclonal antibody specifically detects DNA ligase I in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.

Specifications

Antigen DNA ligase I
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen His-tagged purified human recombinant DNA ligase I overexpressed in a baculovirus/insect cell system.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less