missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DNA Damage ELISA Kit

Catalog No. 89333688
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89333688 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89333688 Supplier Abnova™ Supplier No. KA5026

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use to measure quantitatively human KIT. Sandwich enzyme immunoassay.

DNA Damage ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of DNA and RNA oxidized guanosine species in fetal extract, serum, saliva, plasma, urine, and tissue culture media samples.

Specifications

Product Type ELISA Kit
For Use With (Application) Quant
Quantity 1 Kit
Storage Requirements Store at 4°C