missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DMXL2 Monoclonal Antibody (K70019_7G11), Invitrogen™

Catalog No. MA556116
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556116 200 μL
MA555736 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556116 Supplier Invitrogen™ Supplier No. MA556116
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KNFVDKINLI ERKLFFTTAN AHFKVGCPVL ALEVLSKIPK VTKTSALSAK KDQPDFISHR MDDVPSHSKA LSDGNGSSGI EWSNVTSSQY DWSQPIVKVD EEPLNLDWGE DHDSALDEEE DDAVGLVMKS TDAREKDKQS DQKASDPNML LTPQEEDDPE GDTEVDVIAE QLKFRACLKI LMTELRTLAT GYEVDGGKLR FQLYNWLEKE IAALHEICNH ESVIKEYSSK TYSKVESDLL DQEEMVDKPD IGSYERHQIE RRRLQAKREH

This gene encodes a protein with 12 WD domains. Proteins with WD domains are involved in many functions including participation in signal transduction pathways. Participation of the encoded protein in regulation of the Notch signaling pathway has been demonstrated in vitro using several human cell lines. A gene encoding a similar protein is located on chromosome 5. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen DMXL2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K70019_7G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene DMXL2
Gene Accession No. Q8TDJ6
Gene Alias 6330586A12; 6430411K14Rik; Dmx like 2; DMXL2; Dmx-like 2; dmX-like protein 2; E130119P06Rik; KIAA0856; mKIAA0856; PEPNS; rabconnectin 3; rabconnectin-3; RC3
Gene Symbols DMXL2
Host Species Mouse
Immunogen Recombinant human DMXL2 (immunogen range is 1871-2140).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less