missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DMP1 ELISA Kit

Catalog No. EHDMP1
Encompass
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHDMP1 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EHDMP1 Supplier Invitrogen™ Supplier No. EHDMP1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

The Human DMP1 ELISA quantitates Hu DMP1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu DMP1. Principle of the method The Human DMP1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Dentin matrix acidic phosphoprotein is an extracellular matrix protein and a member of the small integrin binding ligand N-linked glycoprotein family. This protein, which is critical for proper mineralization of bone and dentin, is present in diverse cells of bone and tooth tissues. The protein contains a large number of acidic domains, multiple phosphorylation sites, a functional arg-gly-asp cell attachment sequence, and a DNA binding domain. In undifferentiated osteoblasts it is primarily a nuclear protein that regulates the expression of osteoblast-specific genes. During osteoblast maturation the protein becomes phosphorylated and is exported to the extracellular matrix, where it orchestrates mineralized matrix formation. Mutations in the gene are known to cause autosomal recessive hypophosphatemia, a disease that manifests as rickets and osteomalacia. The gene structure is conserved in mammals. Two transcript variants encoding different isoforms have been described for this gene.

Specifications

Accession Number Q13316
Assay Range 80-40000 pg/mL
Assay Sensitivity 80 pg/mL
Calibration Range 80 pg/mL to 40000 pg/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA antibody pair detect rhDMP-1, it shows no cross-reactivity with rmDMP-1, rhCD44, or rhComplement Factor H
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AG1; ARHP; ARHR; AV020965; dentin matrix acidic phosphoprotein 1; dentin matrix protein 1; DENTMAT; Dmp; DMP 1; Dmp1; DMP-1; MGC130441; PP; serine rich acidic phosphoproteinAG1; ARHP; ARHR; AV020965; dentin matrix acidic phosphoprotein 1; dentin matrix protein 1; DENTMAT; Dmp; DMP 1; Dmp1; DMP-1; MGC130441; PP; serine rich acidic phosphoprotein;
Gene ID (Entrez) 1758
Gene Symbol Dmp1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases DMP-1, Dentin matrix protein 1
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym ARHP, ARHR, DMP-1
Target DMP1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less