missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DMAP1 Recombinant Rabbit Monoclonal Antibody (23GB5680)
GREENER_CHOICE

Catalog No. MA553098
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553098 100 μL
MA553099 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553098 Supplier Invitrogen™ Supplier No. MA553098
Only null left

Rabbit Recombinant Monoclonal Antibody

DMAP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

DNA methyltransferase 1-associated protein 1 (DMAP1) is involved in the repression or activation of transcription. The encoded protein can independently repress transcription and is targeted to replication foci throughout S phase by interacting directly with the N-terminus of DNA methyltransferase 1. During late S phase, histone deacetylase 2 is added to this complex, providing a means to deacetylate histones in transcriptionally inactive heterochromatin following replication. The protein also specifically potentiates DAXX-mediated repression of glucocorticoid receptor-dependent transcription. The encoded protein is also a component of the nucleosome acetyltransferase of H4 complex and interacts with the transcriptional corepressor tumor susceptibility gene 101 and the pro-apoptotic death-associated protein 6, among others. DMAP1 is found in a wide variety of living organisms from yeasts to humans. The human DMAP1 gene is localized on chromosome 1p34.

Specifications

Antigen DMAP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5680
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DMAP1
Gene Accession No. Q9JI44, Q9NPF5
Gene Alias 1500016M21Rik; DKFZp686L09142; Dmap1; Dmtap1; DNA methyltransferase 1 associated protein 1; DNA methyltransferase 1-associated protein 1; DNMAP1; DNMT1 associated protein 1; DNMT1 associated protein-1; DNMT1-associated protein 1; DNMTAP1; EAF2; FLJ11543; KIAA1425; MAT1-mediated transcriptional repressor; MEAF2; mKIAA1425; Mmtr; RP5-891H21.2; SWC4
Gene Symbols DMAP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DMAP1 (411-467AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298447, 55929, 66233
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less