missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DLG5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122034
Description
DLG5 Polyclonal Antibody for Western Blot
May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.Specifications
| DLG5 | |
| Polyclonal | |
| Unconjugated | |
| Dlg5 | |
| 4933429D20Rik; discs large homolog 5; discs large MAGUK scaffold protein 5; discs large protein LP-DLG; discs large protein P-dlg; discs, large homolog 5; discs, large homolog 5 (Drosophila); disks large homolog 5; DLG5; KIAA0583; large type of P-DLG; LP-DLG; mKIAA0583; PDLG; P-DLG5; Placenta and prostate DLG; T25557 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 71228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| E9Q9R9 | |
| Dlg5 | |
| Recombinant protein Discs, Large Homolog 5. The antigen corresponds to amino acid range 1724-1917 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |