missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DLG5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5119087

 View more versions of this product

Catalog No. PIPA5119087


Only null left
Explore available promotions

Description

Description

DLG5 Polyclonal Antibody for Western Blot

May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.
Specifications

Specifications

DLG5
Polyclonal
Unconjugated
Dlg5
4933429D20Rik; discs large homolog 5; discs large MAGUK scaffold protein 5; discs large protein LP-DLG; discs large protein P-dlg; discs, large homolog 5; discs, large homolog 5 (Drosophila); disks large homolog 5; DLG5; KIAA0583; large type of P-DLG; LP-DLG; mKIAA0583; PDLG; P-DLG5; Placenta and prostate DLG; T25557
Rabbit
Antigen affinity chromatography, Protein A
RUO
71228
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
E9Q9R9
Dlg5
Recombinant protein Discs, Large Homolog 5. The antigen corresponds to amino acid range 1724-1917 of the target protein.
200 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions