missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Valine, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 452830010
| Quantity | 1kg |
|---|---|
| Packaging | Plastic bottle |
Chemical Identifiers
| C5H11NO2 | |
| MFCD00004267 | |
| dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid | |
| CHEBI:27266 | |
| CC(C)C(N)C(O)=O |
Specifications
| DL-Valine | |
| C5H11NO2 | |
| MFCD00004267 | |
| KZSNJWFQEVHDMF-UHFFFAOYNA-N | |
| 2-amino-3-methylbutanoic acid | |
| 1kg | |
| CHEBI:27266 | |
| Extra Pure |
| 516-06-3 | |
| Plastic bottle | |
| dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid | |
| CC(C)C(N)C(O)=O | |
| 117.15 | |
| 1182 | |
| 117.15 |