missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| DL-Tyrosine | |
| White-Yellow | |
| MFCD00063074 | |
| OUYCCCASQSFEME-UHFFFAOYNA-N | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 181.19 | |
| CHEBI:18186 | |
| ≥98.0% (HPLC,T) |
| 556-03-6 | |
| C9H11NO3 | |
| dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
| NC(CC1=CC=C(O)C=C1)C(O)=O | |
| 25 g | |
| 1153 | |
| 181.19 | |
| Crystalline Powder |
Chemical Identifiers
| 556-03-6 | |
| 181.19 | |
| OUYCCCASQSFEME-UHFFFAOYNA-N | |
| 1153 | |
| 2-amino-3-(4-hydroxyphenyl)propanoic acid |
| C9H11NO3 | |
| MFCD00063074 | |
| dl-tyrosine, h-dl-tyr-oh, 2-amino-3-4-hydroxyphenyl propanoic acid, tyrosin, tyrosine, dl, l-tyrosine, free base, tirosina, l-tryosine, 3-4-hydroxyphenyl-dl-alanine, benzenepropanoic acid, s | |
| CHEBI:18186 | |
| NC(CC1=CC=C(O)C=C1)C(O)=O |
Safety and Handling
EINECSNumber : (9)-1596
TSCA : Yes