missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Tryptophan, 98%

CAS: 54-12-6 | C11H12N2O2 | 204.23 g/mol

$144.83 - $144.83

Chemical Identifiers

CAS 54-12-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064339
InChI Key QIVBCDIJIAJPQS-UHFFFAOYSA-N
Synonym dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
PubChem CID 1148
ChEBI CHEBI:27897
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=CC=C12)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172110250
SDS View Documents
Thermo Scientific Chemicals
172110250
25 g Plastic Bottle
Each for $144.83
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54-12-6
204.23
QIVBCDIJIAJPQS-UHFFFAOYSA-N
1148
2-amino-3-(1H-indol-3-yl)propanoic acid
C11H12N2O2
MFCD00064339
dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
CHEBI:27897
NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

54-12-6
Authentic
98%
C11H12N2O2
MFCD00064339
dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
NC(CC1=CNC2=CC=CC=C12)C(O)=O
25 g
1148
204.23
Crystalline Powder
Beige to White or Yellow
0.8% max. (105°C, 3 hrs) (vacuum)
(TLC) Not detected
Plastic Bottle
22, 550
QIVBCDIJIAJPQS-UHFFFAOYSA-N
2-amino-3-(1H-indol-3-yl)propanoic acid
204.23
CHEBI:27897
≥97.5%
DL-Tryptophan, 99%
Videos
SDS
Documents

Documents

RUO – Research Use Only