missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Threonine (contains DL-Allothreonine) 97.0+%, TCI America™
Supplier: TCI America T3105100G
Specifications
| 230°C | |
| 80-68-2 | |
| C4H9NO3 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| C[C@@H](O)[C@@H](N)C(O)=O | |
| 100 g | |
| 205 | |
| 119.12 | |
| Crystalline Powder |
| DL-Threonine (contains DL-Allothreonine) | |
| White | |
| MFCD00063722 | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| (2R,3R)-2-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:38263 | |
| ≥97.0% (T) |
Chemical Identifiers
| 80-68-2 | |
| 119.12 | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| 205 | |
| (2R,3R)-2-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| MFCD00063722 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| CHEBI:38263 | |
| C[C@@H](O)[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : (9)-1584
TSCA : Yes