missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Threonine (contains DL-Allothreonine) 97.0+%, TCI America™
$72.70 - $197.59
Chemical Identifiers
| CAS | 80-68-2 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| MDL Number | MFCD00063722 |
| InChI Key | AYFVYJQAPQTCCC-PWNYCUMCSA-N |
| Synonym | dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l |
| PubChem CID | 205 |
| ChEBI | CHEBI:38263 |
| IUPAC Name | (2R,3R)-2-amino-3-hydroxybutanoic acid |
| SMILES | C[C@@H](O)[C@@H](N)C(O)=O |
Chemical Identifiers
| 80-68-2 | |
| 119.12 | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| 205 | |
| (2R,3R)-2-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| MFCD00063722 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| CHEBI:38263 | |
| C[C@@H](O)[C@@H](N)C(O)=O |
Specifications
| 230°C | |
| White | |
| MFCD00063722 | |
| AYFVYJQAPQTCCC-PWNYCUMCSA-N | |
| (2R,3R)-2-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:38263 | |
| ≥97.0% (T) | |
| DL-Threonine (contains DL-Allothreonine) |
| 80-68-2 | |
| C4H9NO3 | |
| dl-threonine, allo-dl-threonine, threonine, dl, dl-allothreonine, dl-2-amino-3-hydroxybutanoic acid, threonine l, h-dl-thr-oh, dl-allo-threonine, allothreonine, d, wln: qy1&yzvq-l | |
| C[C@@H](O)[C@@H](N)C(O)=O | |
| 25 g | |
| 205 | |
| 119.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1584
TSCA : Yes