missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Thioctic Acid, 98+%

Catalog No. AC138720250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138720250 25 g Glass bottle
AC138720050 5 g Glass bottle
AC138721000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138720250 Supplier Thermo Scientific Chemicals Supplier No. 138720250
Only null left

CAS: 1077-28-7 | C8H14O2S2 | 206.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1

Specifications

Melting Point 60°C to 63°C
Color Yellow
Boiling Point 160°C to 165°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9479
Solubility Information (2% in 10% NaOH) Clear yellow
Formula Weight 206.32
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Thioctic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-071-2

RUO – Research Use Only