accessibility menu, dialog, popup

UserName

HOOSS

CAS RN 1077-28-7

IUPAC Name: 5-(1,2-dithiolan-3-yl)pentanoic acid
Synonyms: 6,8-thioctic acid α-liponsaeure thioctansaeure liponic acid 6,8-thiotic acid acetate-replacing factor biletan thioctsaeure 5-(dithiolan-3-yl)valeric acid 6-thioctic acid
Molecular Weight (g/mol): 206.32
Molecular Formula: C8H14O2S2
InChi Key: AGBQKNBQESQNJD-UHFFFAOYNA-N
SMILES: OC(=O)CCCCC1CCSS1
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)