missing translation for 'onlineSavingsMsg'
Learn More

DL-tert-Leucine, 95%

Catalog No. p-7054635
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6300703 1 g
AAH6300706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6300703 Supplier Thermo Scientific Chemicals Supplier No. H6300703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33105-81-6 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33105-81-6
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00065933
InChI Key NPDBDJFLKKQMCM-UHFFFAOYSA-N
Synonym dl-tert-leucine, dl-tert-butylglycine, tert-butylglycine, h-dl-tle-oh, tert-leucine, 3-methylvaline, 2-amino-3,3-dimethyl-butyric acid, 3-methyl valine, t-butylglycine, dl-t-butylglycine
PubChem CID 306131
ChEBI CHEBI:72770
IUPAC Name 2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Specifications

Melting Point >300°C (decomposition)
Beilstein 1721823
Quantity 1 g
Formula Weight 131.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material DL-tert-Leucine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only