accessibility menu, dialog, popup

UserName

H3CCH3H3CNH3OHO

CAS RN 33105-81-6

IUPAC Name: 2-amino-3,3-dimethylbutanoic acid
Synonyms: tert-butylglycine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: NPDBDJFLKKQMCM-UHFFFAOYSA-N
SMILES: CC(C)(C)C(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)