missing translation for 'onlineSavingsMsg'
Learn More

DL-Tartaric Acid 99.0+%, TCI America™

Catalog No. T000125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T000125G 25 g
T0001500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T000125G Supplier TCI America Supplier No. T000125G

Chemical Identifiers

CAS 133-37-9
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00071626
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
PubChem CID 875
ChEBI CHEBI:15674
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 199°C
Color White
Formula Weight 150.09
Percent Purity ≥99.0% (T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material DL-Tartaric Acid
EINECSNumber (2)-1456
TSCA Yes