Learn More
DL-Tartaric acid, 99%
CAS: 133-37-9 | C4H6O6 | 150.09 g/mol
Supplier: Thermo Scientific Chemicals A106830B
Description
DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.
Solubility
Soluble in water, alcohol, mineral acids and alkalies.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
| DL-Tartaric acid | |
| 133-37-9 | |
| MFCD00071626 | |
| 14,9069 | |
| Soluble in water,alcohol,mineral acids and alkalies. | |
| OC(C(O)C(O)=O)C(O)=O | |
| 150.09 | |
| CHEBI:15674 | |
| 99% |
| 210°C to 212°C (decomposition) | |
| C4H6O6 | |
| 1725148 | |
| dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 2,3-dihydroxybutanedioic acid | |
| 875 | |
| 150.09 | |
| 1000 g |
Chemical Identifiers
| 133-37-9 | |
| 150.09 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 875 | |
| 2,3-dihydroxybutanedioic acid |
| C4H6O6 | |
| MFCD00071626 | |
| dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid | |
| CHEBI:15674 | |
| OC(C(O)C(O)=O)C(O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-105-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only