missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich DL-Tartaric acid

Catalog No. T400500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T400500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T400500G Supplier Sigma Aldrich Supplier No. T400500G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DL-Tartaric acid

Specifications

Melting Point 210°C to 212°C (lit.)
CAS 133-37-9
Molecular Formula C4H6O6
Linear Formula HOOC(CHOH)2 COOH
MDL Number MFCD00071626
Synonym DL-2,3-Dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
Percent Purity 99%
Quantity 500 g