missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Serine Methyl Ester Hydrochloride 98.0+%, TCI America™
$32.28 - $49.36
Chemical Identifiers
| CAS | 5619-04-5 |
|---|---|
| Molecular Formula | C4H10ClNO3 |
| Molecular Weight (g/mol) | 155.58 |
| MDL Number | MFCD00012593 |
| InChI Key | NDBQJIBNNUJNHA-UHFFFAOYNA-N |
| Synonym | dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908 |
| PubChem CID | 517373 |
| IUPAC Name | methyl 2-amino-3-hydroxypropanoate;hydrochloride |
| SMILES | COC(=O)C(CO)N.Cl |
Chemical Identifiers
| C4H10ClNO3 | |
| MFCD00012593 | |
| dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908 | |
| methyl 2-amino-3-hydroxypropanoate;hydrochloride |
Specifications
| 136°C | |
| White-Yellow | |
| MFCD00012593 | |
| NDBQJIBNNUJNHA-UHFFFAOYNA-N | |
| methyl 2-amino-3-hydroxypropanoate;hydrochloride | |
| 517373 | |
| ≥98.0% (N,T) | |
| DL-Serine Methyl Ester Hydrochloride |
| 5619-04-5 | |
| C4H10ClNO3 | |
| dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908 | |
| COC(=O)C(CO)N.Cl | |
| 155.58 | |
| 155.58 | |
| Crystalline Powder |
Safety and Handling
TSCA : No