missing translation for 'onlineSavingsMsg'
Learn More

DL-Serine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. S05765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05765G 5 g
S057625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05765G Supplier TCI America Supplier No. S05765G

Chemical Identifiers

CAS 5619-04-5
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
MDL Number MFCD00012593
InChI Key NDBQJIBNNUJNHA-UHFFFAOYNA-N
Synonym dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
PubChem CID 517373
IUPAC Name methyl 2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 155.58
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Serine Methyl Ester Hydrochloride