missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Serine, 99%
CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol
$119.10 - $419.58
Chemical Identifiers
| CAS | 302-84-1 |
|---|---|
| Molecular Formula | C3H7NO3 |
| Molecular Weight (g/mol) | 105.093 |
| MDL Number | MFCD00064223 |
| InChI Key | MTCFGRXMJLQNBG-UHFFFAOYSA-N |
| Synonym | dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid |
| PubChem CID | 617 |
| ChEBI | CHEBI:17822 |
| IUPAC Name | 2-amino-3-hydroxypropanoic acid |
| SMILES | C(C(C(=O)O)N)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1518422
|
Thermo Scientific Chemicals
A1518422 |
100 g |
Each for $119.10
|
|
|||||
|
AAA1518436
|
Thermo Scientific Chemicals
A1518436 |
500 g |
Each for $419.58
|
|
|||||
Description
DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
| C3H7NO3 | |
| MFCD00064223 | |
| dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
| CHEBI:17822 | |
| C(C(C(=O)O)N)O |
Specifications
| ∼240°C (decomposition) | |
| C3H7NO3 | |
| 1721402 | |
| dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
| MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
| 2-amino-3-hydroxypropanoic acid | |
| 617 | |
| 105.09 | |
| DL-Serine |
| 302-84-1 | |
| MFCD00064223 | |
| 14,8460 | |
| Soluble in water. | |
| C(C(C(=O)O)N)O | |
| 105.093 | |
| CHEBI:17822 | |
| 99% |
Safety and Handling
EINECSNumber : 206-130-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only