missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Serine, 99%

Catalog No. AAA1518422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1518422 100 g
AAA1518436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1518422 Supplier Thermo Scientific Chemicals Supplier No. A1518422

CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol

DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 302-84-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064223
InChI Key MTCFGRXMJLQNBG-UHFFFAOYSA-N
Synonym dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid
PubChem CID 617
ChEBI CHEBI:17822
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 1721402
Merck Index 14,8460
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 105.09
Percent Purity 99%
Chemical Name or Material DL-Serine
EINECSNumber 206-130-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only