missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Serine, ≥98%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210286825
Specifications
DL-Serine | |
≥98% | |
3-hydroxyalanine, dl-2-amino-3-hydroxypropionic acid, dl-serine, h-d-ser-oh, h-dl-ser-oh, serin, serine dl-form, serine, dl, serine, l | |
C(C(C(=O)O)N)O | |
105.093 | |
CHEBI:17822 | |
Crystalline Powder | |
White |
302-84-1 | |
C3H7NO3 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
2-amino-3-hydroxypropanoic acid | |
617 | |
105.1 | |
1.537g/mL | |
25g |
Chemical Identifiers
302-84-1 | |
105.093 | |
3-hydroxyalanine, dl-2-amino-3-hydroxypropionic acid, dl-serine, h-d-ser-oh, h-dl-ser-oh, serin, serine dl-form, serine, dl, serine, l | |
CHEBI:17822 | |
C(C(C(=O)O)N)O |
C3H7NO3 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
617 | |
2-amino-3-hydroxypropanoic acid |
Safety and Handling
EINECSNumber : 206-130-6
Recommended Storage : Store at room temperature.