missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Serine, ≥98%, MP Biomedicals™
$98.00
Chemical Identifiers
| CAS | 302-84-1 |
|---|---|
| Molecular Formula | C3H7NO3 |
| Molecular Weight (g/mol) | 105.093 |
| InChI Key | MTCFGRXMJLQNBG-UHFFFAOYSA-N |
| Synonym | dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid |
| PubChem CID | 617 |
| ChEBI | CHEBI:17822 |
| IUPAC Name | 2-amino-3-hydroxypropanoic acid |
| SMILES | C(C(C(=O)O)N)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10286825
|
N/A | 25 g |
Each for $98.00
|
|
|||||
Chemical Identifiers
| 302-84-1 | |
| 105.093 | |
| dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
| CHEBI:17822 | |
| C(C(C(=O)O)N)O |
| C3H7NO3 | |
| MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
| 617 | |
| 2-amino-3-hydroxypropanoic acid |
Specifications
| 302-84-1 | |
| ≥98% | |
| dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
| C(C(C(=O)O)N)O | |
| 2-amino-3-hydroxypropanoic acid | |
| 617 | |
| 105.1 | |
| DL-Serine |
| White | |
| C3H7NO3 | |
| MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
| 1.537g/mL | |
| 105.093 | |
| CHEBI:17822 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 206-130-6
Recommended Storage : Store at room temperature.