missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Selenomethionine 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| UN Number | 3283 |
| Formula Weight | 196.11 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-Selenomethionine |
Safety and Handling
| RTECSNumber | ES7110000 |
| TSCA | No |
| Recommended Storage | Refrigerator |