accessibility menu, dialog, popup

UserName

CAS RN 1464-42-2

SeNH2OHOH3C

CAS RN 1464-42-2

IUPAC Name: 2-amino-4-(methylselanyl)butanoic acid
Synonyms: (+,-)-selenomethionine|selenomethionine, (+,-)-|(+-)-selenomethionine|selenomethionine|SeMet|L(+)-selenomethionine|seleno-L-methionine|selenium methionine|seleno-DL-methionine|methionine, seleno
Molecular Weight (g/mol): 196.12
Molecular Formula: C5H11NO2Se
InChi Key: RJFAYQIBOAGBLC-UHFFFAOYNA-N
SMILES: C[Se]CCC(N)C(O)=O
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)