Learn More
Thermo Scientific Chemicals DL-Selenomethionine, 99+%
CAS: 1464-42-2 | C5H11NO2Se | 196.12 g/mol
$308.77 - $977.47
Chemical Identifiers
| CAS | 1464-42-2 |
|---|---|
| Molecular Formula | C5H11NO2Se |
| Molecular Weight (g/mol) | 196.12 |
| MDL Number | MFCD00063089 |
| InChI Key | RJFAYQIBOAGBLC-UHFFFAOYNA-N |
| Synonym | selenomethionine, dl-selenomethionine, seleno-dl-methionine, +--selenomethionine, butanoic acid, 2-amino-4-methylseleno, selenium methionine, methionine, seleno, selenomethionine van, 2-amino-4-methylselanyl butanoic acid, seleno-d,l-methionine |
| PubChem CID | 15103 |
| ChEBI | CHEBI:27585 |
| IUPAC Name | 2-amino-4-methylselanylbutanoic acid |
| SMILES | C[Se]CCC(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC240775000
|
Thermo Scientific Chemicals
240775000 |
500 mg | Glass bottle |
Each for $308.77
|
|
||||
|
AC240770010
|
Thermo Scientific Chemicals
240770010 |
1 g | Glass bottle |
Each for $539.02
|
|
||||
|
AC240770025
|
Thermo Scientific Chemicals
240770025 |
2.5 g | Glass bottle |
Each for $977.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1464-42-2 | |
| 196.12 | |
| RJFAYQIBOAGBLC-UHFFFAOYNA-N | |
| 15103 | |
| 2-amino-4-methylselanylbutanoic acid |
| C5H11NO2Se | |
| MFCD00063089 | |
| selenomethionine, dl-selenomethionine, seleno-dl-methionine, +--selenomethionine, butanoic acid, 2-amino-4-methylseleno, selenium methionine, methionine, seleno, selenomethionine van, 2-amino-4-methylselanyl butanoic acid, seleno-d,l-methionine | |
| CHEBI:27585 | |
| C[Se]CCC(N)C(O)=O |
Specifications
| 1464-42-2 | |
| Colorless | |
| 99% min. (HPLC) | |
| Glass bottle | |
| CH3SeCH2CH2CH(NH2)COOH | |
| 500 mg | |
| selenomethionine, dl-selenomethionine, seleno-dl-methionine, +--selenomethionine, butanoic acid, 2-amino-4-methylseleno, selenium methionine, methionine, seleno, selenomethionine van, 2-amino-4-methylselanyl butanoic acid, seleno-d,l-methionine | |
| RJFAYQIBOAGBLC-UHFFFAOYNA-N | |
| 2-amino-4-methylselanylbutanoic acid | |
| 15103 | |
| 196.11 | |
| Glistening Powder |
| 265.0°C | |
| Authentic | |
| Faint Smelling | |
| C5H11NO2Se | |
| MFCD00063089 | |
| 15, 8578 | |
| Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in benzene, diethyl ether | |
| C[Se]CCC(N)C(O)=O | |
| 196.12 | |
| CHEBI:27585 | |
| 99+% | |
| DL-Selenomethionine, 99+% |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 215-977-
RTECSNumber : ES7110000
RUO – Research Use Only