missing translation for 'onlineSavingsMsg'
Learn More

DL-Propargylglycine, 96%

Catalog No. p-4228882
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC437320050 Supplier Thermo Scientific Chemicals Supplier No. 437320050

CAS: 64165-64-6 | C5H7NO2 | 113.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64165-64-6
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00056728
InChI Key DGYHPLMPMRKMPD-UHFFFAOYNA-N
Synonym dl-propargylglycine, 2-amino-4-pentynoic acid, h-dl-pra-oh, 2-azanylpent-4-ynoic acid, dl-2-propynylglycine, 2-propynylglycine, l-2-amino-4-pentynoic acid, d,l-propargylglycine, propargyl glycine, +--2-amino-4-pentynoic acid
PubChem CID 95575
IUPAC Name 2-aminopent-4-ynoic acid
SMILES NC(CC#C)C(O)=O

Specifications

Melting Point 235.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 113.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-Propargylglycine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only