CAS RN 64165-64-6
IUPAC Name:
2-aminopent-4-ynoic acid
Synonyms:
propargylglycine
Molecular Weight (g/mol):
113.12
Molecular Formula:
C5H7NO2
InChi Key:
DGYHPLMPMRKMPD-UHFFFAOYNA-N
SMILES:
NC(CC#C)C(O)=O
1
–
1
of
1
results
1
Promotions Available
DL-Propargylglycine, 96%
DL-Propargylglycine, 96%
CAS: 64165-64-6 Molecular Formula: C5H7NO2 Molecular Weight (g/mol): 113.12 MDL Number: MFCD00056728 InChI Key: DGYHPLMPMRKMPD-UHFFFAOYNA-N Synonym: dl-propargylglycine,2-amino-4-pentynoic acid,h-dl-pra-oh,2-azanylpent-4-ynoic acid,dl-2-propynylglycine,2-propynylglycine,l-2-amino-4-pentynoic acid,d,l-propargylglycine,propargyl glycine,+--2-amino-4-pentynoic acid PubChem CID: 95575 IUPAC Name: 2-aminopent-4-ynoic acid SMILES: NC(CC#C)C(O)=O