missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| DL-Prolinol | |
| Yellow | |
| C5H11NO | |
| 25 g | |
| HVVNJUAVDAZWCB-UHFFFAOYSA-N | |
| pyrrolidin-2-ylmethanol | |
| 90908 | |
| 101.15 | |
| Liquid |
| 498-63-5 | |
| 76°C | |
| MFCD00601073 | |
| 2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol # | |
| C1CC(NC1)CO | |
| 101.149 | |
| CHEBI:59652 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 498-63-5 | |
| 101.149 | |
| HVVNJUAVDAZWCB-UHFFFAOYSA-N | |
| 90908 | |
| pyrrolidin-2-ylmethanol |
| C5H11NO | |
| MFCD00601073 | |
| 2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol # | |
| CHEBI:59652 | |
| C1CC(NC1)CO |
Safety and Handling
TSCA : No