missing translation for 'onlineSavingsMsg'
Learn More

DL-Prolinol 95.0+%, TCI America™

Catalog No. p-7135257
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P17935G 5 g
P179325G 25 g
2 options

Catalog No. P17935G

Supplier: TCI America P17935G

Only null left

Chemical Identifiers

CAS 498-63-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00601073
InChI Key HVVNJUAVDAZWCB-UHFFFAOYSA-N
Synonym 2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol #
PubChem CID 90908
ChEBI CHEBI:59652
IUPAC Name pyrrolidin-2-ylmethanol
SMILES C1CC(NC1)CO

Specifications

Color Yellow
Boiling Point 76°C
Quantity 5 g
Formula Weight 101.15
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material DL-Prolinol