missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Proline 99.0+%, TCI America™
$46.20 - $213.69
Chemical Identifiers
| CAS | 609-36-9 |
|---|---|
| Molecular Formula | C5H9NO2 |
| Molecular Weight (g/mol) | 115.132 |
| MDL Number | MFCD00005250 |
| InChI Key | ONIBWKKTOPOVIA-UHFFFAOYSA-N |
| Synonym | dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 |
| PubChem CID | 614 |
| ChEBI | CHEBI:26271 |
| IUPAC Name | pyrrolidine-2-carboxylic acid |
| SMILES | C1CC(NC1)C(=O)O |
Chemical Identifiers
| 609-36-9 | |
| 115.132 | |
| ONIBWKKTOPOVIA-UHFFFAOYSA-N | |
| 614 | |
| pyrrolidine-2-carboxylic acid |
| C5H9NO2 | |
| MFCD00005250 | |
| dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 | |
| CHEBI:26271 | |
| C1CC(NC1)C(=O)O |
Specifications
| 609-36-9 | |
| C5H9NO2 | |
| dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275 | |
| C1CC(NC1)C(=O)O | |
| 1 g | |
| 614 | |
| 115.13 | |
| Crystalline Powder |
| White | |
| MFCD00005250 | |
| ONIBWKKTOPOVIA-UHFFFAOYSA-N | |
| pyrrolidine-2-carboxylic acid | |
| 115.132 | |
| CHEBI:26271 | |
| ≥99.0% (T) | |
| DL-Proline |
Safety and Handling
EINECSNumber : (9)-1626
TSCA : Yes