missing translation for 'onlineSavingsMsg'
Learn More

DL-Proline, 99%, Thermo Scientific Chemicals

$78.92 - $1230.42

Chemical Identifiers

CAS 609-36-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00005250
InChI Key ONIBWKKTOPOVIA-UHFFFAOYSA-N
Synonym dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
PubChem CID 614
ChEBI CHEBI:26271
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1313706
View Documents
Thermo Scientific Chemicals
A1313706
5 g
Each for $78.92
Only null left
 
AAA1313714
View Documents
Thermo Scientific Chemicals
A1313714
25 g
Each for $365.42
Only null left
 
AAA1313722
View Documents
Thermo Scientific Chemicals
A1313722
100 g
Each for $1,230.42
Only null left
 
Description

Description

DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Proline is used as an agonist of glycine receptors. It acts as a catalyst for aldol condensation. It is an essential component of collagen. It plays a significant role in the immune system and strengthening of heart muscles. It is an osmoprotectant and therefore is used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in water and absolute alcohol. Insoluble in ether, butanol and isopropanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

609-36-9
115.132
ONIBWKKTOPOVIA-UHFFFAOYSA-N
614
pyrrolidine-2-carboxylic acid
C5H9NO2
MFCD00005250
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
CHEBI:26271
C1CC(NC1)C(=O)O

Specifications

∼209°C (decomposition)
C5H9NO2
80809
dl-proline, proline, h-dl-pro-oh, proline, dl, 2-pyrrolidine carboxylic acid, hpro, l-proline, homopolymer, +/--pyrrolidine-2-carboxylic acid, prolin, chembl72275
ONIBWKKTOPOVIA-UHFFFAOYSA-N
pyrrolidine-2-carboxylic acid
115.132
CHEBI:26271
99%
609-36-9
MFCD00005250
14,7780
Soluble in water and absolute alcohol. Insoluble in ether,butanol and isopropanol.
C1CC(NC1)C(=O)O
5 g
614
115.13
DL-Proline
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-189-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only