missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-alpha-Phenylglycine, 99%

Catalog No. p-3758672
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130641000 100 g Plastic bottle
AC130645000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC130641000 Supplier Thermo Scientific Chemicals Supplier No. 130641000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2835-06-5 | C8H9NO2 | 151.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2835-06-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00064402
InChI Key ZGUNAGUHMKGQNY-UHFFFAOYNA-N
Synonym DL-α-Aminophenylacetic acid
IUPAC Name 2-amino-2-phenylacetic acid
SMILES NC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 290°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH(NH2)CO2H
Quantity 100 g
Beilstein 14, 460
Merck Index 15, 7402
Specific Rotation -1° to +1° (20°C, 589nm) (c=1, 1N HCl)
Formula Weight 151.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-α-Phenylglycine
Show More Show Less
EINECSNumber 220-608-1

RUO – Research Use Only