accessibility menu, dialog, popup

UserName

CAS RN 2835-06-5

IUPAC Name: 2-amino-2-phenylacetic acid
Synonyms: (+/-)-α-phenylglycine DL-phenylglycine α-phenylgycine α-aminobenzeneacetate α-amino-α-toluate α-amino-α-toluic acid α-toluic acid, α-amino- α-aminobenzeneacetic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: ZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)